Learn More
Med Vet International Braun Catgut 50m Cassette Suture, Size 3, J0091C3
Catalog No. 5000581141
Encompass_Preferred
Manufactured under continuous supervision from selectedmaterials processed to high quality standards. It is accurately gauged and evenly chromed. High knot-pull tensile strength. Good knot-holding ability and exceptionally good handling qualities. Sterilized. 3 USP, 50m (164') L
Catalog No. 50-005-81141 Supplier Med Vet International Supplier No. JOR91C3
Manufactured under continuous supervision from selectedmaterials processed to high quality standards. It is accurately gauged and evenly chromed. High knot-pull tensile strength. Good knot-holding ability and exceptionally good handling qualities. Sterilized. 3 USP, 50m (164') L