Learn More
Med Vet International Braun Empty IV Bag, 150ml, J1318C, Each
Catalog No. 5000581117
Encompass_Preferred
150ml capacity. Sterile Partial Additive Bag (PAB) - Latex free - PVC free - DEHP free - Dual ports: Add port and port for admin- istration set - Hanging loop on top Rigid port allows safer and easier addition of prescribed drugs. No over wraps: ready to use.
Catalog No. 50-005-81117 Supplier Med Vet International Supplier No. JOR1318C
150ml capacity. Sterile Partial Additive Bag (PAB) - Latex free - PVC free - DEHP free - Dual ports: Add port and port for admin- istration set - Hanging loop on top Rigid port allows safer and easier addition of prescribed drugs. No over wraps: ready to use.