Learn More
Med Vet International Braun Stainless Steel Suture Cassette, #0, 50M, J0091S0
Catalog No. 5000583401
Encompass_Preferred
#0 USP 50m (164').Monofilament, corrosion- resistant steel. Extremely high knotpull tensile strength. Stitch holds for an unlimited period of time. Sterilized.
Catalog No. 50-005-83401 Supplier Med Vet International Supplier No. JOR0091S0
#0 USP 50m (164').Monofilament, corrosion- resistant steel. Extremely high knotpull tensile strength. Stitch holds for an unlimited period of time. Sterilized.