Learn More
Med Vet International Braun Stainless Steel Suture Cassette, #2, 50M, J0091S20
Catalog No. 5000583179
Encompass_Preferred
Size #2 USP, 50m (164'). Monofilament, corrosion- resistant steel. Extremely high knotpull tensile strength. Stitch holds for an unlimited period of time. Sterilized.
Catalog No. 50-005-83179 Supplier Med Vet International Supplier No. JOR0091S20
Size #2 USP, 50m (164'). Monofilament, corrosion- resistant steel. Extremely high knotpull tensile strength. Stitch holds for an unlimited period of time. Sterilized.