Learn More
Med Vet International Braunamid Cassette Suture, Size 0 USP, Heavy 75m, J009100
Catalog No. 5000583614
Encompass_Preferred
Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than monofil sutures. Knot-pull tensile strength substantially exceeds USP requirements. Knot-holding ability is excellent due to its inner polyfilament structure. Non-capillary. Sterilized. Braunamid meets or exceeds all legal requirements set forth by the U.S. government.
Catalog No. 50-005-83614 Supplier Med Vet International Supplier No. JOR009100
Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than monofil sutures. Knot-pull tensile strength substantially exceeds USP requirements. Knot-holding ability is excellent due to its inner polyfilament structure. Non-capillary. Sterilized. Braunamid meets or exceeds all legal requirements set forth by the U.S. government.