Learn More
Med Vet International Braunamid Stainless Steel Suture Cassette, #2, 25M, J0091S2
Catalog No. 5000583362
Encompass_Preferred
#2 USP, 25m (82'). Monofilament, corrosion- resistant steel. Extremely high knotpull tensile strength. Stitch holds for an unlimited period of time. Sterilized.
Catalog No. 50-005-83362 Supplier Med Vet International Supplier No. JOR0091S2
#2 USP, 25m (82'). Monofilament, corrosion- resistant steel. Extremely high knotpull tensile strength. Stitch holds for an unlimited period of time. Sterilized.