Learn More
Med Vet International Braunamid With Needle 2/0, 3/8in Reverse Cutting Needle, 45cm, DS30B20, 12 Count
Catalog No. 5000582442
Encompass_Preferred
3/8-Circle Reverse Cutting Needle; 45cm SIZE #2/0 The triangular, reverse cutting needle is intended for suturing tough tissue. The three edges run over the entire length of the needle, and give the needle holder a firm grip.
Catalog No. 50-005-82442 Supplier Med Vet International Supplier No. JORS30B20
3/8-Circle Reverse Cutting Needle; 45cm SIZE #2/0 The triangular, reverse cutting needle is intended for suturing tough tissue. The three edges run over the entire length of the needle, and give the needle holder a firm grip.