Learn More
Fluotics Bravo comp P30 non-sterile Type A (1536 Plate accessible)
Catalog No. NC2195895
Encompass_Preferred
Agilent Bravo compatible P30, non-sterile, Type A (1536 Plate accessible), working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 11484-302
This item is not returnable. View return policy
Catalog No. NC2195895 Supplier Fluotics Supplier No. AGI30.NS/A
Agilent Bravo compatible P30, non-sterile, Type A (1536 Plate accessible), working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 11484-302