Learn More
Fluotics Bravo comp P70 sterile wide bore
Supplier:  Fluotics AGI70WB.ST
Encompass_Preferred
Agilent Bravo compatible P70, sterile, wide bore, working volume: 70 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 19134-012
Catalog No. NC2195908