Learn More
Anatech Ltd BRAZILLIANT 1QT
Catalog No. NC9982671
Encompass_Preferred
BRAZILLIANT,1 Qt,Aqueous soln of Brazilin,use:A red nuclear stain in histopathology,Component:Acetic acid,Aluminum ammonium sulfate,Brazilin,Reagent alcohol,Sodium iodate,pH:2.4-2.7,Physical state:Liquid,Boiling range:212degreeF,Specific gr
Catalog No. NC9982671 Supplier Anatech Ltd Supplier No. 861
BRAZILLIANT,1 Qt,Aqueous soln of Brazilin,use:A red nuclear stain in histopathology,Component:Acetic acid,Aluminum ammonium sulfate,Brazilin,Reagent alcohol,Sodium iodate,pH:2.4-2.7,Physical state:Liquid,Boiling range:212degreeF,Specific gr