Learn More
Cayman Chemical BRC4wt (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502918795
Encompass_Preferred
An inhibitor of the BRCA2-RAD51 protein-protein interaction; shortens DNA replication tract lengths and reduces homologous repair frequency of camptothecin-induced DNA damage in vitro, as well as potentiates olaparib-induced cell death in HeLa and U2OS cells but not in non-cancerous hTERT RPE-1, MRC-5, or MCF-10A cells when coupled to (Arg)9
Catalog No. 50-291-8795 Supplier Cayman Chemical Supplier No. 3884810 MG
An inhibitor of the BRCA2-RAD51 protein-protein interaction; shortens DNA replication tract lengths and reduces homologous repair frequency of camptothecin-induced DNA damage in vitro, as well as potentiates olaparib-induced cell death in HeLa and U2OS cells but not in non-cancerous hTERT RPE-1, MRC-5, or MCF-10A cells when coupled to (Arg)9