Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 672MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: BRCA1/1398. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: A431 or HeLa Cells. Breast, Ovarian or Renal Cell Carcinoma. Cellular Staining: Cytoplasmic and Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human BRCA1 protein fragment (aa445-620). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 220kDa.Chromosome: 17q21. Gene Symbol: BRCA1. Gene ID: 672. Ensemble: ENSG00000012048. Uniprot: NP_009229.
Catalog No. 50-188-5193