Learn More
Medchemexpress LLC HY-100862 5mg Medchemexpress, BRCA1-IN-2 CAS:1622262-55-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008625MG
Encompass_Preferred
Medchemexpress, HY-100862 5mg BRCA1-IN-2 CAS:1622262-55-8 BRCA1-IN-2 (compound 15) is a cell-permeable protein-protein interaction (PPI) inhibitor for BRCA1 with an IC50 of 0.31 μM and a Kd of 0.3 μM, which shows antitumor activities via the disruption of BRCA1 (BRCT)2/protein interactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0606