Learn More
Aviva Systems Biology Corp BRCC3 Antibody - C-terminal region (ARP61990_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRCC3. It was validated on Western Blot. . Alternate names: BRCC36, C6.1A, CXorf53, RP11-143H17.2ApplicationIF, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-532