Learn More
Medchemexpress LLC HY-112719 5mg Medchemexpress, BRD 4354 CAS:315698-07-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127195MG
Encompass_Preferred
Medchemexpress, HY-112719 5mg BRD 4354 CAS:315698-07-8 BRD 4354 is a moderately potent inhibitor of HDAC5 and HDAC9 , with IC 50 s of 0.85 and 1.88 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2434