Learn More
Medchemexpress LLC HY-112060 10mg Medchemexpress, Saccharin 1-methylimidazole CAS:482333-74-4 Purity:>98%
SDP
Catalog No. 501969006
Encompass_Preferred
Medchemexpress, HY-112719B 10mg BRD 4354 (ditrifluoroacetate) CAS: BRD 4354 (ditrifluoroacetate) is a moderately potent inhibitor of HDAC5 and HDAC9, with IC50s of 0.85 and 1.88 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9006 Supplier Medchemexpress LLC Supplier No. HY11206010MG
Medchemexpress, HY-112719B 10mg BRD 4354 (ditrifluoroacetate) CAS: BRD 4354 (ditrifluoroacetate) is a moderately potent inhibitor of HDAC5 and HDAC9, with IC50s of 0.85 and 1.88 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.