Learn More
Medchemexpress LLC HY-100719 5mg Medchemexpress, BRD-6929 CAS:849234-64-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007195MG
Encompass_Preferred
Medchemexpress, HY-100719 5mg BRD-6929 CAS:849234-64-6 BRD-6929 (Cpd-60) is a brain-penetrant, selective inhibitor of HDAC1 and HDAC2 (IC50= 1 and 8 nM), extracted from patent US2018360927. BRD-6929 (Cpd-60) shows high-affinity to HDAC1 and HDAC2 with Ki of 0.2 and 1.5 nM, respectively. BRD-6929 (TPB) potentiates the efficacy of gnidimacrin (a PKC Agonist) against latent HIV-1[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7775