Learn More
Medchemexpress LLC BRD-6929 | 849234-64-6 | MFCD16495786 | 99.5% | 351.4 g/mol | C19H17N3O2S | 25 MG
SDP
Catalog No. 5000213687
Encompass_Preferred
BRD-6929 is a potent, selective, brain-penetrant inhibitor of class I histone deacetylases HDAC1 and HDAC2, showing low-nanomolar inhibitory activity and high binding affinity. It is used in preclinical research, including CNS and mood-related behavioral model studies, for target validation and pharmacology characterization.
  • Potent HDAC1 and HDAC2 inhibition (IC50 1 nM and 8 nM).
  • High binding affinity to targets (Ki 0.2 nM and 1.5 nM).
  • Brain-penetrant properties suitable for CNS research.
  • Applicable to mood-related behavioral model studies and target validation.
  • Supplied in small research quantities for preclinical experiments.
Catalog No. 50-002-13687 Supplier Medchemexpress LLC Supplier No. HY10071925MG
BRD-6929 is a potent, selective, brain-penetrant inhibitor of class I histone deacetylases HDAC1 and HDAC2, showing low-nanomolar inhibitory activity and high binding affinity. It is used in preclinical research, including CNS and mood-related behavioral model studies, for target validation and pharmacology characterization.
  • Potent HDAC1 and HDAC2 inhibition (IC50 1 nM and 8 nM).
  • High binding affinity to targets (Ki 0.2 nM and 1.5 nM).
  • Brain-penetrant properties suitable for CNS research.
  • Applicable to mood-related behavioral model studies and target validation.
  • Supplied in small research quantities for preclinical experiments.