Learn More
Apexbio Technology LLC BRD 7552 1137359-47-7 50mg
SDP
Catalog No. 5000087669
Encompass_Preferred
BRD 7552 is a small-molecule inducer targeting pancreatic duodenal homeobox-1 (PDX1) a transcription factor involved in pancreatic development and -cell function It is designed to enhance PDX1 expression thereby promoting -cell gene regulatory pathways BRD7552 exerts its biological activity primarily through epigenetic modulation of the PDX1 promoter region increasing both mRNA and protein expression levels In cell-based studies BRD7552 demonstrates concentration-dependent induction of insulin gene expression in human PANC-1 ductal cells and primary human islet cultures Based on these pharmacological properties BRD7552 holds research potential in studies of pancreatic cell transdifferentiation -cell function mechanisms and diabetes therapy development strategies
Catalog No. 50-000-87669 Supplier Apexbio Technology LLC Supplier No. B778250
BRD 7552 is a small-molecule inducer targeting pancreatic duodenal homeobox-1 (PDX1) a transcription factor involved in pancreatic development and -cell function It is designed to enhance PDX1 expression thereby promoting -cell gene regulatory pathways BRD7552 exerts its biological activity primarily through epigenetic modulation of the PDX1 promoter region increasing both mRNA and protein expression levels In cell-based studies BRD7552 demonstrates concentration-dependent induction of insulin gene expression in human PANC-1 ductal cells and primary human islet cultures Based on these pharmacological properties BRD7552 holds research potential in studies of pancreatic cell transdifferentiation -cell function mechanisms and diabetes therapy development strategies