Learn More
TARGETMOL CHEMICALS INC BRD 9757 5MG
SDP
Catalog No. 502580764
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. BRD 9757 (N-Hydroxy-1-cyclopentene-1-carboxamide) is a selective inhibitor of HDAC6 (IC50 = 30 nM). purity: 99%
Catalog No. 50-258-0764 Supplier TARGETMOL CHEMICALS INC Supplier No. T251755MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. BRD 9757 (N-Hydroxy-1-cyclopentene-1-carboxamide) is a selective inhibitor of HDAC6 (IC50 = 30 nM). purity: 99%