Learn More
Medchemexpress LLC HY-114268 1mg Medchemexpress, BRD-K98645985 CAS:1357647-78-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1142681MG
Encompass_Preferred
Medchemexpress, HY-114268 1mg BRD-K98645985 CAS:1357647-78-9 BRD-K98645985 is a BAF (mammalian SWI/SNF) transcriptional repression inhibitor with an EC50 of ~2.37 µM. BRD-K98645985 binds ARID1A-specific BAF complexes, prevents nucleosomal positioning, and potently reverses HIV-1 latency, without T cell activation or toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9782