Learn More
Grainger Hose, PVC Braid, Clear, 1.025" OD, 3/4", 100 ft, Roll
Catalog No. NC2736736
Encompass_Preferred
The FARPVC1712-100P is a 100 foot roll of 1/4" ID, 3/8" OD clear PVC braided reinforced hose from Flex-A-Prene. This flexible hose is made of PVC with a polyester braid reinforcement and has a pressure rating of 150 PSI. It has a temperature range of -20°F to 150°F. The inside diameter is 1/4" and outside diameter is 3/8". This general purpose hose is suitable for air, water and many chemicals. The hose is sold by Grainger in a 100 foot roll.
Catalog No. NC2736736 Supplier Grainger Supplier No. 742T24
The FARPVC1712-100P is a 100 foot roll of 1/4" ID, 3/8" OD clear PVC braided reinforced hose from Flex-A-Prene. This flexible hose is made of PVC with a polyester braid reinforcement and has a pressure rating of 150 PSI. It has a temperature range of -20°F to 150°F. The inside diameter is 1/4" and outside diameter is 3/8". This general purpose hose is suitable for air, water and many chemicals. The hose is sold by Grainger in a 100 foot roll.