Learn More
Medchemexpress LLC HY-116830 10mg Medchemexpress, BRD0705 CAS:2056261-41-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11683010MG
Encompass_Preferred
Medchemexpress, HY-116830 10mg BRD0705 CAS:2056261-41-5 BRD0705 is a potent, paralog selective and orally active GSK3α inhibitor with an IC50 of 66 nM and a Kd of 4.8 μM. BRD0705 displays increased selectivity for GSK3α (8-fold) versus GSK3β (IC50 of 515 nM). BRD0705 can be used for acute myeloid leukemia (AML). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9443