Learn More
Aviva Systems Biology Corp BRD2 Antibody - middle region (ARP34212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BRD2. Alternate names: FSH, NAT, RNF3, FSRG1, RING3, D6S113EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-55302