Learn More
Cayman Chemical BRD2 bromodomaIn 2humnre 100ug
SDP
Catalog No. 502951088
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged BRD2 bromodomain 2 expressed in E. coli • AA: 339-459 • MW: 42.4 kDa
Catalog No. 50-295-1088 Supplier Cayman Chemical Supplier No. 11070100 UG
Source: Recombinant N-terminal GST-tagged BRD2 bromodomain 2 expressed in E. coli • AA: 339-459 • MW: 42.4 kDa