Learn More
Cayman Chemical BRD2 bromodomaIns 1 nd 2hu
SDP
Catalog No. 502951200
Encompass_Preferred
Source: Recombinant GST-tagged protein expressed in E. coli • Amino acids: 65-459 • MW: 71.2 kDa
Catalog No. 50-295-1200 Supplier Cayman Chemical Supplier No. 11069100 UG
Source: Recombinant GST-tagged protein expressed in E. coli • Amino acids: 65-459 • MW: 71.2 kDa