Learn More
Aviva Systems Biology Corp BRD3 Antibody - middle region (ARP38940_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38940_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BRD3. Alternate names: KIAA0043, ORFX, RING3LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56856