Learn More
Cayman Chemical BRD3 bromodomaIn 2humnre 100ug
SDP
Catalog No. 502971063
Encompass_Preferred
Source: Recombinant human N-terminal GST-tagged BRD3 bromodomain 2 expressed in E. coli • Amino acids: 306-416 • MW: 40.7 kDa
Catalog No. 50-297-1063 Supplier Cayman Chemical Supplier No. 14658100 UG
Source: Recombinant human N-terminal GST-tagged BRD3 bromodomain 2 expressed in E. coli • Amino acids: 306-416 • MW: 40.7 kDa