Learn More
Cayman Chemical BRD3 bromodomaIns 1 nd 2hu
SDP
Catalog No. 502971455
Encompass_Preferred
Source: Recombinant human N-terminal GST-tagged BRD3 bromodomains 1 and 2 expressed in E. coli • Amino acids: 2-434 • MW: 75 kDa
Catalog No. 50-297-1455 Supplier Cayman Chemical Supplier No. 14864100 UG
Source: Recombinant human N-terminal GST-tagged BRD3 bromodomains 1 and 2 expressed in E. coli • Amino acids: 2-434 • MW: 75 kDa