Learn More
Medchemexpress LLC HY-19618 5mg Medchemexpress, BRD3308 CAS:1550053-02-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196185MG
Encompass_Preferred
Medchemexpress, HY-19618 5mg BRD3308 CAS:1550053-02-5 BRD3308 is a highly selective HDAC3 inhibitor with an IC50 of 54 nM. BRD3308 is 23-fold selectivity for HDAC3 over HDAC1 (IC50 of 1.26 μM) or HDAC2 (IC50 of 1.34 μM). BRD3308 suppresses pancreatic β-cell apoptosis induced by inflammatory cytokines or glucolipotoxic stress, and increases functional insulin release. BRD3308 activates HIV-1 transcription and disrupts HIV-1 latency[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0864