Learn More
Medchemexpress LLC BRD3731 (S enantiomer) | 2056262-07-6 | 98.2% | 377.52 g/mol | C24H31N3O | 100MG
SDP
Catalog No. 5000236375
Encompass_Preferred
BRD3731 (S-enantiomer) is a selective GSK3β inhibitor supplied for preclinical biochemical and cell-based research. It has been reported as a GSK3β-selective compound (from patent US20160375006A1) and is used in studies of mood disorders, metabolic disease, and neurodegeneration. The compound is provided as a solid or as DMSO solution formats and is characterized by defined physicochemical properties and high purity.
Catalog No. 50-002-36375 Supplier Medchemexpress LLC Supplier No. HY124607B100MG
BRD3731 (S-enantiomer) is a selective GSK3β inhibitor supplied for preclinical biochemical and cell-based research. It has been reported as a GSK3β-selective compound (from patent US20160375006A1) and is used in studies of mood disorders, metabolic disease, and neurodegeneration. The compound is provided as a solid or as DMSO solution formats and is characterized by defined physicochemical properties and high purity.