Learn More
Santa Cruz Biotechnology BRD4 Antibody (A-7) HRP, 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC518021 HRP
Encompass
BRD4 Antibody (A-7) is recommended for detection of BRD4 of human origin by WB, IP, IF and ELISA. HRP is the preferred detection reagent for BRD4 Antibody (A-7) for WB applications.
Catalog No. NC1823596