Learn More
Sino Biological Recombinant Human PADI3/PAD3 Protein (Full Length, GST Tag), Active
Catalog No. 502573755
Encompass_Preferred
Recombinant full length human PAD3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3755 Supplier Sino Biological Supplier No. P312310CG50
Recombinant full length human PAD3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.