Learn More
Cayman Chemical BRD4 bromodomaIn 2humnre 100ug
SDP
Catalog No. 502952386
Encompass_Preferred
Source: Recombinant N-terminal His-tagged BRD4 bromodomain 2 expressed in E. coli • Amino acids: 342-460 (partial length) • MW: 15.8 kDa
Catalog No. 50-295-2386 Supplier Cayman Chemical Supplier No. 11721100 UG
Source: Recombinant N-terminal His-tagged BRD4 bromodomain 2 expressed in E. coli • Amino acids: 342-460 (partial length) • MW: 15.8 kDa