Learn More
Cayman Chemical BRD4 bromodomaIns 1 nd 2hu
SDP
Supplier:  Cayman Chemical 14822100 UG
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged BRD4 bromodomains 1 and 2 expressed in E. coli • Amino acids: 2-477 (partial protein) • MW: 80.6 kDa
Catalog No. 50-297-0694