Learn More
Cayman Chemical BRD4 bromodomaIns 1 nd 2hu
SDP
Catalog No. 502951320
Encompass_Preferred
Source: Recombinant human N-terminal GST-tagged BRD4 bromodomains 1 and 2 expressed in E. coli • Amino acids: 49-460 • MW: 73.4 kDa
Catalog No. 50-295-1320 Supplier Cayman Chemical Supplier No. 11052100 UG
Source: Recombinant human N-terminal GST-tagged BRD4 bromodomains 1 and 2 expressed in E. coli • Amino acids: 49-460 • MW: 73.4 kDa