Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reaction Biology Corp BRD4 Full Length protein, human recombinant, 100 μg, N-terminal His-tag, C-terminal StrepII-tag
SDP

Catalog No. NC2889035
Encompass

The REACTION BIOLOGY CORP BRD4 Full Length protein is a human recombinant bromodomain-containing protein 4 that plays a significant role in transcriptional regulation and cell cycle progression. This protein, produced in Sf9 insect cells, features N-terminal His and C-terminal StrepII tags, enabling specific binding to ε-N-acetyllysine residues. Mainly recognized for its function in chromatin regulation, cancer research, and inflammation studies, BRD4 is also an atypical kinase that phosphorylates RNA Polymerase II.

  • Produced in Sf9 insect cells
  • Active transcriptional regulator
  • Contains two bromodomains
  • Has a molecular weight of 156.5 kDa
  • Tagged with N-terminal His and C-terminal StrepII
  • Purity exceeds 85% by SDS-PAGE
  • Stored at -70°C or -80°C
  • Functionally important for BET bromodomain studies

Catalog No. NC2889035 Supplier Reaction Biology Corp Supplier No. RD21153100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The REACTION BIOLOGY CORP BRD4 Full Length protein is a human recombinant bromodomain-containing protein 4 that plays a significant role in transcriptional regulation and cell cycle progression. This protein, produced in Sf9 insect cells, features N-terminal His and C-terminal StrepII tags, enabling specific binding to ε-N-acetyllysine residues. Mainly recognized for its function in chromatin regulation, cancer research, and inflammation studies, BRD4 is also an atypical kinase that phosphorylates RNA Polymerase II.

  • Produced in Sf9 insect cells
  • Active transcriptional regulator
  • Contains two bromodomains
  • Has a molecular weight of 156.5 kDa
  • Tagged with N-terminal His and C-terminal StrepII
  • Purity exceeds 85% by SDS-PAGE
  • Stored at -70°C or -80°C
  • Functionally important for BET bromodomain studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.