Learn More
Medchemexpress LLC BRD4 inhibitor-10 | 1660117-38-3 | 99.5% | 429.51 | C25H27N5O2 | 10 MG
SDP
Catalog No. 5000228275
Encompass_Preferred
BRD4 Inhibitor-10 is a potent BRD4-BD1 inhibitor (Compound II-25) used in epigenetic and oncology research. It has a reported IC50 of 8 nM, a molecular weight of 429.51 g·mol⁻¹, and high reported purity for laboratory studies.
  • Potent BRD4-BD1 inhibitor with IC50 8 nM.
  • High reported purity (99.52%).
  • Available as solid and as DMSO solution formats for flexible use.
  • Suitable for epigenetic, cellular proliferation, and cancer biology research.
Catalog No. 50-002-28275 Supplier Medchemexpress LLC Supplier No. HY11749110MG
BRD4 Inhibitor-10 is a potent BRD4-BD1 inhibitor (Compound II-25) used in epigenetic and oncology research. It has a reported IC50 of 8 nM, a molecular weight of 429.51 g·mol⁻¹, and high reported purity for laboratory studies.
  • Potent BRD4-BD1 inhibitor with IC50 8 nM.
  • High reported purity (99.52%).
  • Available as solid and as DMSO solution formats for flexible use.
  • Suitable for epigenetic, cellular proliferation, and cancer biology research.