Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRD4 Inhibitor-10 | 1660117-38-3 | 99.5% | 429.51 g·mol⁻¹ | C25H27N5O2 | 50 MG
SDP

Catalog No. 5000228177
Encompass_Preferred

BRD4 Inhibitor-10 is a potent BRD4-BD1 inhibitor (reported IC50 8 nM) supplied as a 50 mg solid for preclinical research in epigenetics and cancer biology.

  • Potent BRD4-BD1 inhibition with reported IC50 of 8 nM.
  • High purity (~99.5%) suitable for biochemical and cellular assays.
  • Molecular weight 429.51 g·mol⁻¹ and formula C25H27N5O2.
  • Solid, off-white to light yellow appearance for easy handling.
  • Recommended storage: powder at -20°C (stable ~3 years) or 4°C (~2 years).

Catalog No. 50-002-28177 Supplier Medchemexpress LLC Supplier No. HY11749150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRD4 Inhibitor-10 is a potent BRD4-BD1 inhibitor (reported IC50 8 nM) supplied as a 50 mg solid for preclinical research in epigenetics and cancer biology.

  • Potent BRD4-BD1 inhibition with reported IC50 of 8 nM.
  • High purity (~99.5%) suitable for biochemical and cellular assays.
  • Molecular weight 429.51 g·mol⁻¹ and formula C25H27N5O2.
  • Solid, off-white to light yellow appearance for easy handling.
  • Recommended storage: powder at -20°C (stable ~3 years) or 4°C (~2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.