Learn More
Medchemexpress LLC BRD4 Inhibitor-24 | 309951-18-6 | 99.27% | 262.26 | 50 MG
SDP
Catalog No. 5000362892
Encompass_Preferred
BRD4 Inhibitor-24 (compound 3U) is a potent BRD4 inhibitor that has demonstrated antitumor activity. It shows IC50 values of 33.7 μM against MCF7 cells and 45.9 μM against K652 cells. This compound has been extracted from patent CN107721975A.
  • Potent BRD4 inhibitor.
  • Exhibits antitumor activity against MCF7 (IC50: 33.7 μM) and K652 (IC50: 45.9 μM) cells.
  • Derived from patent CN107721975A.
  • For research use only.
Catalog No. 50-003-62892 Supplier Medchemexpress LLC Supplier No. HY14720250MG
BRD4 Inhibitor-24 (compound 3U) is a potent BRD4 inhibitor that has demonstrated antitumor activity. It shows IC50 values of 33.7 μM against MCF7 cells and 45.9 μM against K652 cells. This compound has been extracted from patent CN107721975A.
  • Potent BRD4 inhibitor.
  • Exhibits antitumor activity against MCF7 (IC50: 33.7 μM) and K652 (IC50: 45.9 μM) cells.
  • Derived from patent CN107721975A.
  • For research use only.