Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRD4 Inhibitor-24 | 309951-18-6 | 99.3% | 262.26 | 100 MG
SDP

Catalog No. 5000363956
Encompass_Preferred

BRD4 Inhibitor-24 (compound 3U) is a potent BRD4 inhibitor that exhibits antitumor activity against MCF7 and K652 cells. This compound is for research use only and is presented as a solid, white to off-white substance with the molecular formula C13H14N2O4.

  • Potent BRD4 inhibitor
  • Antitumor activity against MCF7 and K652 cells (IC50 values of 33.7 and 45.9 μM)
  • Solid, white to off-white appearance
  • Molecular formula C13H14N2O4
  • Recommended storage at 4°C, protected from light
  • In solvent, storage at -80°C for 6 months or -20°C for 1 month (protected from light)

Catalog No. 50-003-63956 Supplier Medchemexpress LLC Supplier No. HY147202100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRD4 Inhibitor-24 (compound 3U) is a potent BRD4 inhibitor that exhibits antitumor activity against MCF7 and K652 cells. This compound is for research use only and is presented as a solid, white to off-white substance with the molecular formula C13H14N2O4.

  • Potent BRD4 inhibitor
  • Antitumor activity against MCF7 and K652 cells (IC50 values of 33.7 and 45.9 μM)
  • Solid, white to off-white appearance
  • Molecular formula C13H14N2O4
  • Recommended storage at 4°C, protected from light
  • In solvent, storage at -80°C for 6 months or -20°C for 1 month (protected from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.