Learn More
Medchemexpress LLC BRD4 inhibitor-24 | 309951-18-6 | 99.3% | C13H14N2O4 | 10MG
SDP
Catalog No. 5000258218
Encompass_Preferred
BRD4 Inhibitor-24 is a high-purity small-molecule inhibitor of the BRD4 bromodomain with reported antitumor activity in MCF7 and K562 cells. Supplied for research use, it is characterized for molecular properties, solubility, and storage to support in vitro and in vivo studies.
  • High purity (99.3%).
  • Molecular formula C13H14N2O4; molecular weight 262.26.
  • Reported antitumor activity in MCF7 and K562 cell lines.
  • High solubility in DMSO (≥100 mg/mL) and suitable in vivo formulation (≥2.5 mg/mL in 10% DMSO, 90% corn oil).
  • Defined storage stability in solution: -80°C for up to 6 months, -20°C for up to 1 month.
Catalog No. 50-002-58218 Supplier Medchemexpress LLC Supplier No. HY14720210MG
BRD4 Inhibitor-24 is a high-purity small-molecule inhibitor of the BRD4 bromodomain with reported antitumor activity in MCF7 and K562 cells. Supplied for research use, it is characterized for molecular properties, solubility, and storage to support in vitro and in vivo studies.
  • High purity (99.3%).
  • Molecular formula C13H14N2O4; molecular weight 262.26.
  • Reported antitumor activity in MCF7 and K562 cell lines.
  • High solubility in DMSO (≥100 mg/mL) and suitable in vivo formulation (≥2.5 mg/mL in 10% DMSO, 90% corn oil).
  • Defined storage stability in solution: -80°C for up to 6 months, -20°C for up to 1 month.