Learn More
Medchemexpress LLC BRD4 Inhibitor-24 (compound 3U) | 309951-18-6 | 99.3% | 262.26 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1472025MG
Encompass_Preferred
BRD4 Inhibitor-24 (compound 3U) is a potent BRD4 inhibitor that demonstrates antitumor activity against MCF7 and K652 cells, with IC50 values of 33.7 and 45.9 μM, respectively. This compound is extracted from patent CN107721975A.
  • Potent BRD4 inhibitor
  • Shows antitumor activity against MCF7 and K652 cells (IC50 values of 33.7 and 45.9 μM)
  • Appears as a solid with a white to off-white color
  • Soluble in DMSO for in vitro use (≥ 100 mg/mL)
  • Suitable for in vivo experiments with 10% DMSO and 90% corn oil (≥ 2.5 mg/mL clear solution)
Catalog No. 50-003-61283