Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRD4 ligand-linker conjugate 1 TFA | 00-00-0 | 98.7% | 614.06 g/mol | C30H31ClF3N7O2 | 5 MG
SDP

Catalog No. 5000262604
Encompass_Preferred

BRD4 ligand-linker conjugate 1 TFA is a trifluoroacetic acid salt ligand-linker conjugate designed as a BRD4-targeting building block for use in the synthesis of PROTACs. It is supplied as a viscous liquid with high purity and is intended for research use in medicinal chemistry and chemical biology.

  • Target protein ligand-linker conjugate for BRD4 used in PROTAC synthesis.
  • Supplied as a viscous liquid, light yellow to brown in color.
  • High purity (~98.7%), suitable for synthetic applications.
  • Molecular weight 614.06 g/mol; molecular formula C30H31ClF3N7O2.
  • Recommended storage: solid at 4°C under nitrogen; in solution store at -80°C for up to 6 months.

Catalog No. 50-002-62604 Supplier Medchemexpress LLC Supplier No. HY132943A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRD4 ligand-linker conjugate 1 TFA is a trifluoroacetic acid salt ligand-linker conjugate designed as a BRD4-targeting building block for use in the synthesis of PROTACs. It is supplied as a viscous liquid with high purity and is intended for research use in medicinal chemistry and chemical biology.

  • Target protein ligand-linker conjugate for BRD4 used in PROTAC synthesis.
  • Supplied as a viscous liquid, light yellow to brown in color.
  • High purity (~98.7%), suitable for synthetic applications.
  • Molecular weight 614.06 g/mol; molecular formula C30H31ClF3N7O2.
  • Recommended storage: solid at 4°C under nitrogen; in solution store at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.