Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRD4 ligand-linker conjugate 1 TFA | 00-00-0 | 98.7% | C30H31ClF3N7O2 | 10MG
SDP

Catalog No. 5000259724
Encompass_Preferred

BRD4 ligand-Linker Conjugate 1 TFA is a target protein ligand-linker conjugate supplied as the trifluoroacetic acid (TFA) salt for use in the synthesis of PROTACs. The material is provided in pre-weighed laboratory quantities and is characterized for purity and identity.

  • Designed for assembly of PROTAC molecules.
  • Provided as the TFA salt for stability and handling.
  • High purity (98.7%) suitable for synthetic applications.
  • Characterized by molecular weight 614.06 and formula C30H31ClF3N7O2.
  • Supplied in multiple pre-weighed pack sizes for medicinal chemistry use.
  • Structural and analytical data available on the product page.

Catalog No. 50-002-59724 Supplier Medchemexpress LLC Supplier No. HY132943A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRD4 ligand-Linker Conjugate 1 TFA is a target protein ligand-linker conjugate supplied as the trifluoroacetic acid (TFA) salt for use in the synthesis of PROTACs. The material is provided in pre-weighed laboratory quantities and is characterized for purity and identity.

  • Designed for assembly of PROTAC molecules.
  • Provided as the TFA salt for stability and handling.
  • High purity (98.7%) suitable for synthetic applications.
  • Characterized by molecular weight 614.06 and formula C30H31ClF3N7O2.
  • Supplied in multiple pre-weighed pack sizes for medicinal chemistry use.
  • Structural and analytical data available on the product page.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.