Learn More
Apexbio Technology LLC BRD4770 1374601-40-7 10mg
SDP
Catalog No. 502240196
Encompass_Preferred
BRD4770 (CAS 1374601-40-7) is a small-molecule inhibitor targeting the histone methyltransferase G9a with an IC50 of 6 3 M In studies using the PANC-1 pancreatic cancer cell line BRD4770 was shown to decrease levels of di- and trimethylated histone H3 lysine 9 (H3K9) through inhibition of G9a activity This reduction in H3K9 methylation is associated with the induction of cellular senescence and cell death as well as suppression of both adherent-dependent and independent cell proliferation BRD4770 serves as a useful probe for exploring the epigenetic regulation of gene expression and cellular senescence in cancer biology research
This item is not returnable. View return policy
Catalog No. 50-224-0196 Supplier Apexbio Technology LLC Supplier No. B483710
BRD4770 (CAS 1374601-40-7) is a small-molecule inhibitor targeting the histone methyltransferase G9a with an IC50 of 6 3 M In studies using the PANC-1 pancreatic cancer cell line BRD4770 was shown to decrease levels of di- and trimethylated histone H3 lysine 9 (H3K9) through inhibition of G9a activity This reduction in H3K9 methylation is associated with the induction of cellular senescence and cell death as well as suppression of both adherent-dependent and independent cell proliferation BRD4770 serves as a useful probe for exploring the epigenetic regulation of gene expression and cellular senescence in cancer biology research