Learn More
Selleck Chemical LLC BRD4770-5mg
SDP
Supplier:  Selleck Chemical LLC S75915MG
Encompass_Preferred
BRD4770 is a histone methyltransferase G9a inhibitor with IC50 of 6.3 μM, and induces cell senescence.
Catalog No. 50-238-9615