Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRD5529 10mM 1mL | 1358488-78-4 | 1 ML
SDP

Catalog No. 5000307938
Encompass_Preferred

Small-molecule CARD9-TRIM62 protein-protein interaction (PPI) inhibitor supplied for laboratory research and provided as a 10 mM solution in DMSO or as solid material. The compound has a molecular weight of 465.54 g/mol and is supplied at high purity for in vitro studies. Not for human therapeutic or diagnostic use.

  • Effective CARD9-TRIM62 PPI inhibitor with reported IC50 around 8.6 μM.
  • Available as a 10 mM solution in DMSO and in multiple solid vial sizes.
  • High purity (≈98.9%) suitable for biochemical and cellular assays.
  • Molecular weight 465.54 g/mol for calculation of dosing and concentrations.
  • Storage recommendations provided for powder and solution to preserve stability.
  • Intended for research use only; not for clinical or diagnostic applications.

Catalog No. 50-003-07938 Supplier Medchemexpress LLC Supplier No. HY11549710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule CARD9-TRIM62 protein-protein interaction (PPI) inhibitor supplied for laboratory research and provided as a 10 mM solution in DMSO or as solid material. The compound has a molecular weight of 465.54 g/mol and is supplied at high purity for in vitro studies. Not for human therapeutic or diagnostic use.

  • Effective CARD9-TRIM62 PPI inhibitor with reported IC50 around 8.6 μM.
  • Available as a 10 mM solution in DMSO and in multiple solid vial sizes.
  • High purity (≈98.9%) suitable for biochemical and cellular assays.
  • Molecular weight 465.54 g/mol for calculation of dosing and concentrations.
  • Storage recommendations provided for powder and solution to preserve stability.
  • Intended for research use only; not for clinical or diagnostic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.