Learn More
Medchemexpress LLC HY-115497 5mg Medchemexpress, BRD5529 CAS:1358488-78-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1154975MG
Encompass_Preferred
Medchemexpress, HY-115497 5mg BRD5529 CAS:1358488-78-4 BRD5529 is a selective CARD9-E3 ubiquitin ligase TRIM62 protein-protein interaction inhibitor with an IC50 of 8.6 μM. BRD5529 directly and selectively binds CARD9, disrupts TRIM62 recruitment, inhibits TRIM62-mediated ubiquitinylation and activation of CARD9, and demonstrates cellular activity and selectivity in CARD9-dependent pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9803