Learn More
Aviva Systems Biology Corp BRD7 Antibody - C-terminal region (ARP39018_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human BRD7. Alternate names: BP75, CELTIX1, NAG4ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-53124